Learn More
Kartell™ Finned Plug for Disposable Tubes
Description
Designed with flexible fins to create a snug fit.
- Made from polyethylene
- Available in different colors
Specifications
Specifications
| Certifications/Compliance | CE-IVD |
| Product Type | Finned Plug |
| Material | Polyethylene |
| Color | Neutral |
| For Use With (Equipment) | 11 to 13 mm Tubes |