Learn More
3M™ Plastic Sweatband
Description
- Replacement plastic sweatband suitable for use with 3M™ Hard Hats and 3M™ G500 Headgear Combinations
- Quick and easy to install
Specifications
Specifications
| Certifications/Compliance | CE Approved |
| Color | Black |
| For Use With (Equipment) | All 3M™ Hard Hats |
| Material | Plastic |
| Product Type | Sweatband |