Learn More
Epredia™ Plastic Slide Folders
Product Code. 12618336 Shop All Epredia Products
Click to view available options
Color:
Gray
White
Yellow
Unit Size:
Pack of 10
Product Code. 12618336
Brand: Epredia™ B1001200YW
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Easily color-code your slides with these plastic slide folders, available in a variety of colors.
  • Hold up to 20 175 x 26mm (3" x 1") slides for convenient storage
  • Transparent top for easy identification of contents
Please Note: Availability may vary by country.
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Device Holds 20 Slides
Length (English) Slide 3 in.
Length (Metric) Slide 75 mm
Material Plastic
Stackable Yes
Width (English) Slide 1 in.
Width (Metric) Slide 26 mm