Description
- Polystyrene, crystal clear
- For aqueous solutions
- |Polystyrene, crystal clear
Specifications
Specifications
| Material | Polystyrene |
| Sterility | Sterile |
| For Use With (Application) | For Training Purposes, in Industrial Labs, During Field Research and Applications Where Pipettes Made of Glass Cannot be Used, e.g. in Fluorine and Silicate Chemistry |
| Color | Translucent |
| Volume (Metric) | 100 mL |
| Type | Single Use Pipette |
| Disposable | Single-use |