Learn More
Buerkle™ Plastic Single-use Pipets
Shop All Bürkle Products
Click to view available options
Volume (Metric):
25 mL
50 mL
100 mL
Unit Size:
Each
Description
- Polystyrene, crystal clear
- For aqueous solutions
- |Polystyrene, crystal clear
Specifications
Specifications
| Material | Polystyrene |
| Sterility | Sterile |
| For Use With (Application) | For Training Purposes, in Industrial Labs, During Field Research and Applications Where Pipettes Made of Glass Cannot be Used |
| Quantity | Each |
| Color | Crystal Clear |
| Volume (Metric) | 50 mL |
| Type | Disposable Pipette |
| Disposable | Single-use |