Learn More
Buerkle™ Plastic Powder Funnels
Product Code. p-8002443 Shop All Bürkle Products
Change view
Click to view available options
Diameter (Metric)Top:
80 mm
100 mm
150 mm
253 mm
350 mm
Diameter (Metric) Stem:
21 mm
22 mm
28 mm
50 mm
65 mm
Unit Size:
Each
Product Code. Diameter (Metric)Top Diameter (Metric) Stem unitSize
11971491 80 mm 21 mm Each
10641153 100 mm 22 mm Each
10020042 150 mm 28 mm Each
10085091 253 mm 50 mm Each
10091862 350 mm 65 mm Each
5 options
Product Code. 11971491
Brand: Buerkle™ 96060080
Only null left
Add to basket
Add to basket
Powder funnel for safe filling of containers. 9606-0240 with fine sieve made of phosphor-bronze (not for use with foodstuffs).
  • Made of PP/PE
  • For safe filling of powders
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 80 mm
Diameter (Metric) Stem 21 mm
Material Polypropylene
For Use With (Application) For Foodstuffs