Learn More
Buerkle™ Plastic Powder Funnels
Product Code. 10085091 Shop All Bürkle Products
Click to view available options
Diameter (Metric)Top:
80 mm
100 mm
150 mm
253 mm
350 mm
Diameter (Metric) Stem:
21 mm
22 mm
28 mm
50 mm
65 mm
Unit Size:
Each
Product Code. 10085091
Brand: Buerkle™ 96060240
Powder funnel for safe filling of containers. 9606-0240 with fine sieve made of phosphor-bronze (not for use with foodstuffs).
  • Made of PP/PE
  • For safe filling of powders
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 253 mm
Diameter (Metric) Stem 50 mm
Material Polyethylene
For Use With (Application) For Foodstuffs