Learn More
Buerkle™ Plastic Powder Funnels
Product Code. 10020042 Shop All Bürkle Products
Change view
Click to view available options
Diameter (Metric)Top:
80 mm
100 mm
150 mm
253 mm
360 mm
Diameter (Metric) Stem:
25 mm
32 mm
50 mm
65 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Diameter (Metric) Stem Unit Size
10020042 150 mm 32 mm Each
11971491 80 mm 25 mm Each
10641153 100 mm 25 mm Each
10085091 253 mm 50 mm Each
10091862 360 mm 65 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10020042 Supplier Buerkle™ Supplier No. 96060150
  • ONLINE EXCLUSIVE PRICE
    75.85 NOK
    74.90 NOK / Pack of 1
    Save 0.95 NOK
    1% Off
Estimated Shipment: 01-12-2026
Powder funnel for safe filling of containers. 9606-0240 with fine sieve made of phosphor-bronze (not for use with foodstuffs).
  • Efficiently fills powder without loss or spillage
  • Versatile, easy to use and quick to set up
  • Robust and durable, good chemical resistance
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 150 mm
Capacity (Metric) 800 mL
Diameter (Metric) Stem 32 mm
Material Polypropylene
Height (Metric) 138 mm
Color Translucent
For Use With (Application) For Foodstuffs, For Universal Use, For Powdery Substances, As Accessory For Filtration
Length (Metric) Stem 22 mm
Capacity (English) 27.05 oz.
Height (English) 5.43 in.
Length (English) Stem 0.87 in.
Product Type Filling Funnel