Learn More
Cytiva Plastic Manifold Light Kit for Ettan™ IPGphor™ 3 Isoelectric Focusing Unit
Product Code. 10299344 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Ettan IPGphor 3 IEF Unit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10299344 Ettan IPGphor 3 IEF Unit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10299344 Supplier Cytiva Supplier No. 11002688
Estimated Shipment: 14-01-2027
Includes
1 Cleaning brush, 2 Electrode assembly, 1 Spirit level, 20 Sample cup, 1 Sample cup insertion tool, 60 Paper electrode wicks, 60 Paper bridge pads, 1 Manifold, 1 Forceps, 1 60mL cleaning solution, User manual
Manifold Light Kit for Ettan™ IPGphor™ 3 Isoelectric Focusing Unit
  • Material: Plastic
  • Uniform thermal characteristics
  • Speed up micropreparative load processing
  • No wetted part
  • Maximum 12 strips
  • No. of gels: 1 to 12
Highly suitable for routine general screening application
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Highly suitable for routine general screening application
For Use With (Equipment) Ettan IPGphor 3 IEF Unit
Type Complete Plastic Manifold Light Kit
Includes 1 Cleaning brush, 2 Electrode assembly, 1 Spirit level, 20 Sample cup, 1 Sample cup insertion tool, 60 Paper electrode wicks, 60 Paper bridge pads, 1 Manifold, 1 Forceps, 1 60mL cleaning solution, User manual
Material Plastic
No. of Gels 1 to 12