Learn More
Microspec™ Plastic Inoculation Loops
Product Code. 16644822
Change view
Click to view available options
Volume:
1 μL
5 μL
10 μL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 250
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume Sterility Unit Size
16644822 5 μL Sterile Pack of 250
16654822 1 μL Non-sterile Pack of 250
16664822 1 μL Sterile Pack of 250
16634822 5 μL Non-sterile Pack of 250
16664782 10 μL Non-sterile Pack of 250
16674782 10 μL Sterile Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16644822 Supplier Microspec™ Supplier No. B5G
  • ONLINE EXCLUSIVE PRICE
    341.00 NOK
    286.74 NOK / Pack of 250
    Save 54.26 NOK
    15% Off
Estimated Shipment: 16-12-2026
Available as aseptic manufactured or ethylene oxide sterilized.
  • Includes 250 Plastic Inoculation Loops
  • Aseptic Manufactured or Ethylene Oxide Sterilized
  • 200 mm length
Specifications
Disposable Single-use
Volume 5 μL
Sterility Sterile
Color Blue
Length (Metric) Handle 200 mm
Quantity 250
Packaging Type Microseal Pouch