Learn More
Buerkle™ General Purpose Funnels PP/PE
Product Code. 10645873 Shop All Bürkle Products
Click to view available options
Diameter (Metric)Top:
40 mm
60 mm
80 mm
100 mm
120 mm
140 mm
160 mm
Diameter (Metric) Stem:
5 mm
8 mm
10 mm
11 mm
13 mm
16 mm
18 mm
Material:
Polyethylene
Polypropylene
Unit Size:
Each
Product Code. 10645873
Brand: Buerkle™ 96020080
Designed for general purpose use. Buerkle™ Plastic Funnels feature a suspension eyelet for safe filling.
  • Suitable for foodstuff
  • Available in polyethylene or polypropylene
  • Wide range of sizes
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 80 mm
Diameter (Metric) Stem 11 mm
Material Polyethylene
For Use With (Application) For Foodstuffs
Includes Suspension eyelet
Product Type Funnel