Learn More
Buerkle™ General Purpose Funnels PP/PE
Product Code. 10205772 Shop All Bürkle Products
Change view
Click to view available options
Diameter (Metric)Top:
40 mm
60 mm
80 mm
100 mm
120 mm
140 mm
160 mm
Diameter (Metric) Stem:
5 mm
8 mm
10 mm
11 mm
13 mm
16 mm
18 mm
Material:
Polyethylene
Polypropylene
Unit Size:
Each
Product Code. Diameter (Metric)Top Diameter (Metric) Stem Material unitSize
10205772 120 mm 13 mm Polypropylene Each
10659480 40 mm 5 mm Polypropylene Each
10347042 60 mm 8 mm Polyethylene Each
10690733 100 mm 10 mm Polypropylene Each
10645873 80 mm 11 mm Polyethylene Each
10260032 140 mm 16 mm Polyethylene Each
10071722 160 mm 18 mm Polyethylene Each
7 options
Product Code. 10205772
Brand: Buerkle™ 96020120
Only null left
Add to basket
Add to basket
Designed for general purpose use. Buerkle™ Plastic Funnels feature a suspension eyelet for safe filling.
  • Suitable for foodstuff
  • Available in polyethylene or polypropylene
  • Wide range of sizes
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 120 mm
Capacity (Metric) 3 ml
Diameter (Metric) Stem 13 mm
Material Polypropylene
For Use With (Application) For Foodstuffs
Includes Suspension eyelet
Product Type Funnel