Learn More
Microspec™ Plastic Culture Needles
Product Code. 15792125
Click to view available options
Sterility:
Aseptically Manufactured
Gamma-irradiated
Quantity:
1000
16000
Unit Size:
Pack of 1000
Pack of 16000
Product Code. 15792125
Brand: Microspec™ F4G
Only null left
Add to basket
Add to basket
Transfer and inoculate living microorganisms
  • Sharp end to facilitate easy stab inoculation
  • Picks up small colonies
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
Specifications
Disposable Single-use
Length (English) 7.9 in.
Length (Metric) 200 mm
Material High-impact Polystyrene
Sterility Gamma-irradiated
Color Blue
Product Type Inoculating Needle
Quantity 1000
Packaging Type Packaged in Microseal Pouches of 20