Learn More
Hecht Karl™ Pear Shaped Centrifuge Tubes ASTM
Description
- Borosilicate glass 3.3
- DIN 51793
- Fused blue graduation marks
- ASTM D 96
- Cylindrical bottom
Specifications
Specifications
| Certifications/Compliance | ASTM D 96, DIN 51793 |
| Capacity (Metric) | 100 mL |
| Material | Borosilicate Glass (3.3) |
| For Use With | Centrifugation |
| Shape | Pear shaped |
| Bottom Shape | Round |
| Graduations | 0.1 mL (0 to 1.5 mL), 0.5 mL (1.5 to 5 mL), 1 mL (5 to 10 mL), 5 mL (10 to 25 mL), 25 mL (25 to 100 mL) |