missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasma Cell Marker Mouse, Clone: SPM310, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio MSM1X-47-P1
This item is not returnable.
View return policy
Specifications
| Plasma Cell Marker | |
| Cytoplasmic | |
| SPM310 | |
| Unconjugated | |
| Mouse | |
| Tonsil or lymph node | |
| 100 μg | |
| Primary | |
| IgG2a |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| Pancreatic cancer-related mucin | |
| Protein A/G Purified | |
| RUO | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?