Learn More
Motic™ CCIS™ EC-H Plan Phase Objective, Negative Phase
Product Code. 13475489
Click to view available options
Magnification Power:
100X
10X
20X
40X
Unit Size:
Each
Product Code. 13475489
Brand: Motic™ 1101001703401
CCIS™ EC-H Plan Phase Objective, Negative Phase is compatible with all BA series microscopes
  • For use with BA Series Microscopes
  • EC-H Plan Phase
  • Negative Phase
  • Magnifications/Numerical Apertures available:
    • 10X/0.25 (WD=17.4mm)
    • 20X/0.45 (WD=0.9mm)
    • 40X/0.65/S (WD=0.5mm)
    • 100X/1.25/S-Oil (WD=0.15mm)
Specifications
Type CCIS, Plan Achromatic Phase Objective, Negative Phase
Field of View EC-H Plan Phase
Focus Distance 0.9 mm WD
For Use With (Equipment) BA Microscope Series
Magnification Power 20X
Quantity 1 Ea.