Learn More
BRAND™ Plain Interior Polypropylene Funnels
Product Code. 10339761 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric)Top:
50mm
75mm
100mm
120 mm
150mm
Height (Metric):
43 mm (Stem)
55 mm stem
77 mm (Stem)
90 mm (Stem)
95 mm (Stem)
Unit Size:
Pack of 12
Pack of 24
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Height (Metric) Unit Size
10339761 120 mm 90 mm (Stem) Pack of 12
11853910 100mm 77 mm (Stem) Pack of 12
15281459 50mm 43 mm (Stem) Pack of 24
15291459 75mm 55 mm stem Pack of 12
10594062 150mm 95 mm (Stem) Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10339761 Supplier BRAND™ Supplier No. 147030
  • ONLINE EXCLUSIVE PRICE
    520.00 NOK
    424.63 NOK / Pack of 12
    Save 95.37 NOK
    18% Off
Estimated Shipment: 08-12-2026
An excellent option with high temperature resistance for general laboratory use.
  • Available in a range of sizes
  • Sold in various quantities
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 120 mm
Diameter (Metric) Stem 14 mm
Material Polypropylene
Height (Metric) 90 mm (Stem)
Autoclavable Autoclavable
Product Type Filling Funnel