missing translation for 'onlineSavingsMsg'
Learn More

PLAGL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25864-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16844425

  • 1415.13 NOK / 20µL
Estimated Shipment: 13-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a C2H2 zinc finger protein with transactivation and DNA-binding activities. It has been shown to have anti-proliferative properties, and thus thought to function as a tumor suppressor. In addition, overexpression of this gene during fetal development is believed to underlie the rare disorder, transient neonatal diabetes mellitus (TNDM). This gene is imprinted, with preferential expression of the paternal allele in many tissues, however, biallelic expression has been noted in peripheral blood leucocytes. A recent study reports that tissue-specific imprinting results from variable utilization of monoallelic and biallelic promoters. Many transcript variants differing in the 5′ UTR and encoding two different isoforms, have been found for this gene.
Specifications

Specifications

PLAGL1
Polyclonal
Unconjugated
PLAGL1
Lost on transformation 1, LOT 1, LOT1, PLAGL1, Tumor supressor ZAC, ZAC, ZAC1, Zinc finger protein PLAGL1
Rabbit
Antigen Affinity Chromatography
RUO
22634, 5325
-20°C
Liquid
Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UM63
PLAGL1
PLAGL1 Fusion Protein Ag23073
20 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers