missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLAC1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17343-1-AP-150UL
This item is not returnable.
View return policy
Description
The function remains unknown.Specifications
| PLAC1L | |
| Polyclonal | |
| Unconjugated | |
| OOSP2 | |
| PLAC1L, placenta 1 like, TMEM122 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 219990, 225922, 690888 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q4FZG8, Q86WS3 | |
| Oosp2 | |
| PLAC1L Fusion Protein Ag11384 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction