missing translation for 'onlineSavingsMsg'
Learn More

PLA2G4D Mouse anti-Human, Clone: 2E2A10, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66752-1-IG-20UL

 View more versions of this product

Product Code. 16868915

  • 1710.33 NOK / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The phospholipase A2 enzyme family, including PLA2G4D, catalyze the hydrolysis of glycerophospholipids at the sn-2 position and then liberate free fatty acids and lysophospholipids.[supplied by OMIM, Jun 2009].
Specifications

Specifications

PLA2G4D
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q86XP0
PLA2G4D
PLA2G4D Fusion Protein Ag4178
20 μL
Primary
Human
Antibody
IgG1
Western Blot
2E2A10
Unconjugated
PLA2G4D
cPLA2 delta, cPLA2delta, Phospholipase A2 group IVD, PLA2G4D
Mouse
Protein G
RUO
283748
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.