missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G4B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55074-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
PLA2G4B transcribes naturally-occurring mRNAs that are co-transcribed products of the neighboring JMJD7 and PLA2G4B genes. Incompletely processed read-through transcripts from these two loci are abundantly expressed in most tissues, but the function of the predicted protein product has not yet been determined.Specifica
PLA2G4B | |
Polyclonal | |
Unconjugated | |
PLA2G4B | |
cPLA2 beta, HsT16992, Phospholipase A2 group IVB, PLA2G4B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100137049 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P0C869 | |
PLA2G4B | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |