missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PLA2G4A (Human) Recombinant Protein
Human PLA2G4A partial ORF ( NP_077734, 1 a.a. - 110 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005321-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSFIDPYQHIIVEHQYSHKFTVVVLRATKVTKGAFGDMLDTPDPYVELFISTTPDSRKRTRHFNNDINPVWNETFEFILDPNQENVLEITLMDANYVMDETLGTATFTVS
Specifications
NP_077734 | |
phospholipase A2, group IVA (cytosolic, calcium-dependent) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PLA2G4A | |
GST |
5321 | |
Wheat germ expression system | |
25 μg | |
MGC126350, PLA2G4, cPLA2-alpha | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |