missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PLA2G4A (Human) Recombinant Protein

Human PLA2G4A partial ORF ( NP_077734, 1 a.a. - 110 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00005321-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189151

  • 4115.00 NOK / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MSFIDPYQHIIVEHQYSHKFTVVVLRATKVTKGAFGDMLDTPDPYVELFISTTPDSRKRTRHFNNDINPVWNETFEFILDPNQENVLEITLMDANYVMDETLGTATFTVS
Specifications

Specifications

NP_077734
phospholipase A2, group IVA (cytosolic, calcium-dependent)
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PLA2G4A
GST
5321
Wheat germ expression system
10 μg
MGC126350, PLA2G4, cPLA2-alpha
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers