missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15843-1-AP-150UL
This item is not returnable.
View return policy
Description
Phospholipase A2 catalyzes the release of fatty acids from glycero-3-phosphocholines. The best known varieties are the digestive enzymes secreted as zymogens by the pancreas of mammals. Sequences of pancreatic PLA2 enzymes from a variety of mammals have been reported. One striking feature of these enzymes is their close homology to venom phospholipases of snakes. Other forms of PLA2 have been isolated from brain, liver, lung, spleen, intestine, macrophages, leukocytes, erythrocytes, inflammatory exudates, chondrocytes, and platelets.Specifications
| PLA2G1B | |
| Polyclonal | |
| Unconjugated | |
| PLA2G1B | |
| Group IB phospholipase A2, Phospholipase A2, PLA2, PLA2A, PLA2G1B, PPLA2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18778, 29526, 5319 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P04054, P04055, Q9Z0Y2 | |
| PLA2G1B | |
| PLA2G1B Fusion Protein Ag8603 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction