Learn More
Tocris Bioscience™ PKI 166 hydrochloride
Potent EGFR-kinase inhibitor
Brand: Tocris Bioscience™ 4941/10
This item is not returnable.
View return policy
Description
PKI 166 hydrochloride is a potent EGFR-kinase inhibitor (IC50 = 0.7 nM). Displays >3000-fold selectivity against a panel of serine/threonine kinases. Reduces metastasis and angiogenesis in a mouse model of pancreatic cancer. Antihypertensive and orally bioavailable.Specifications
| >98% | |
| 2230253-82-2 | |
| 10 mg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only