Learn More
Tocris Bioscience™ PKI 166 hydrochloride
GREENER_CHOICE
Potent EGFR-kinase inhibitor
Brand:  Tocris Bioscience™ 4941/10
Encompass
Product Code. 15796595
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
PKI 166 hydrochloride is a potent EGFR-kinase inhibitor (IC50 = 0.7 nM). Displays >3000-fold selectivity against a panel of serine/threonine kinases. Reduces metastasis and angiogenesis in a mouse model of pancreatic cancer. Antihypertensive and orally bioavailable.
Specifications
Specifications
>98%
2230253-82-2