Learn More
PKC Alpha Mouse anti-Human, CoraLite 594, Clone: 1D11F4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66421-100UL
Description
The PKC Alpha antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PKC Alpha. This antibody recognizes human, mouse, rat antigen. The PKC Alpha antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| PKC Alpha | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 5578 | |
| IgG1 |
| Immunofluorescence | |
| 1D11F4 | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |