missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PJA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17687-1-AP-20UL
This item is not returnable.
View return policy
Description
Has E2-dependent E3 ubiquitin-protein ligase activity. Ubiquitinates MAGED1 antigen leading to its subsequent degradation by proteasome. May be involved in protein sorting.Specifications
| PJA1 | |
| Polyclonal | |
| Unconjugated | |
| PJA1 | |
| PJA1, praja ring finger 1, Praja1, RING finger protein 70, RNF70 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18744, 64219, 683077 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O55176, Q8NG27 | |
| Pja1 | |
| PJA1 Fusion Protein Ag11820 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction