Learn More
Pivmecillinam hydrochloride, MedChemExpress
Product Code. 16478466
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. Quantity unitSize
16478466 100 mg 100mg
16464497 50 mg 50mg
16408638 10 mg 10mg
3 options
Product Code. 16478466
Brand: Medchem Express HYB0810A
MedChemExpress Pivmecillinam hydrochloride (FL-1039 hydrochloride) is an orally active prodrug of mecillinam, an extended-spectrum penicillin antibiotic.
Pivmecillinam hydrochloride (FL-1039 hydrochloride) is an orally active prodrug of mecillinam, an extended-spectrum penicillin antibiotic.
Specifications
Chemical Name or Material Pivmecillinam hydrochloride
Physical Form Solid
CAS 32887-03-9
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H34ClN3O5S
For Use With (Application) COVID-19-immunoregulation
Synonym FL-1039 hydrochloride
Solubility Information H2O : 100 mg/mL (210.07 mM; Need ultrasonic) ∣DMSO : ≥ 34 mg/mL (71.42 mM)
SMILES O=C([C@@H](C(C)(C)S[C@]1([H])[C@@H]2/N=C/N3CCCCCC3)N1C2=O)OCOC(C(C)(C)C)=O.[H]Cl
Molecular Weight (g/mol) 476.03
Formula Weight 476.03
Percent Purity 98.0%
Grade Research