Learn More
Piston Syphon Pump with Polyethylene Suction Tube
Click to view available options
For Use With (Equipment):
Up to 205 L Drums
Unit Size:
Each
Description
A selection of polyethylene pumps. They can be used either as a pump or in syphoning mode. For safely dispensing liquids from containers ranging from 5 L up to 205 L drums
Specifications
Specifications
| Quantity | 1 Each |
| Type | Piston Syphon Pump |
| For Use With (Application) | For safely dispensing liquids from containers ranging from 5 L up to 205 L drums. |
| For Use With (Equipment) | Up to 205 L Drums |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.