Learn More
PISD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16401-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to PISD. The sequence corresponds to the beta isoform.
Phosphatidylserine decarboxylases catalyze the formation of phosphatidylethanolamine (PE) by decarboxylation of phosphatidylserine (PS). Type I PSDs, such as PISD, are targeted to the inner mitochondrial membrane by an N-terminal targeting sequence. PISD also contains a conserved LGST motif that functions as an autocatalytic cleavage site where the proenzyme is split into mature alpha and beta subunits.Specifications
PISD | |
Polyclonal | |
Unconjugated | |
PISD | |
DJ858B16, PISD, PSDC, PSSC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23761, 320951, 681361 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D3ZAW2, Q8BSF4, Q9UG56 | |
Pisd | |
PISD Fusion Protein Ag9499 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |