Learn More
Piroxicam, TRC
Specifications
Specifications
| Chemical Name or Material | Piroxicam |
| CAS | 36322-90-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 H13 N3 O4 S |
| Quantity | 500 mg |
| Synonym | 4-Hydroxy-2-methyl-N-(2-pyridyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide, Artroxicam, Baxo, Bruxicam, CHF 1251, CP 16171, Caliment, Dispercam, Doblexan, Dolibid, Erazon, Felden gel, Feldene, Feldene Melt, Feldene Zydis, Feldoral, Feledene, Feloxin, Felvin 20, Felxicam, Flogobene, Flogosan, Geldene, Improntal, Larapam, Laxidene, Maxvin, Murupe, NSC 666076, Neoxicam, Nesprex-DT, Nkoyo piroxicam, Pirkam, Piroflex, Piroftal, Piroxicam, Piroxidine, Piroxitas-DT, Pixicam, Pyroxycam, Reudene, Rheugesic, Riacen, Roxicam, Roxiden, Sasulen, Solocalm, Uphaxicam, Zunden |
| InChI Formula | InChI=1S/C15H13N3O4S/c1-18-13(15(20)17-12-8-4-5-9-16-12)14(19)10-6-2-3-7-11(10)23(18,21)22/h2-9,19H,1H3,(H,16,17,20) |
| SMILES | CN1C(=C(O)c2ccccc2S1(=O)=O)C(=O)Nc3ccccn3 |
| IUPAC Name | 4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-yl-1λ^{6},2-benzothiazine-3-carboxamide |
| Molecular Weight (g/mol) | 331.35 |
| Show More |