Learn More
Piracetam, MedChemExpress
Product Code. 16472239
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472239 1 g 1g
16411018 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16472239 Supplier Medchem Express Supplier No. HYB0585
MedChemExpress Piracetam (UCB-6215) is a cyclic derivative of the neurotransmitter gamma-aminobutyric acid (GABA), used in treatment of a wide range of cognitive disorders.
Piracetam (UCB-6215) is a cyclic derivative of the neurotransmitter gamma-aminobutyric acid (GABA), used in treatment of a wide range of cognitive disorders.
Specifications
Chemical Name or Material Piracetam
CAS 7491-74-9
Color White
Purity Grade Notes Research
Molecular Formula C6H10N2O2
Quantity 1 g
Synonym UCB-6215
Solubility Information DMSO : ≥ 100 mg/mL (703.43 mM) ∣H2O : ≥ 50 mg/mL (351.72 mM)
SMILES O=C(N)CN1C(CCC1)=O
Molecular Weight (g/mol) 142.16
Formula Weight 142.16
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration