Learn More
PIR Mouse anti-Human, Unconjugated, Clone: 2A10E4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67516-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16800196
  • 1611.84 NOK / 20µL
Estimated Shipment: 27-05-2024
Description
The PIR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PIR. This antibody recognizes human, mouse, rat antigen. The PIR antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PIR
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
2A10E4
Unconjugated
20 μL
8544
IgG1