Learn More
PIR Mouse anti-Human, Unconjugated, Clone: 2A10E4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67516-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The PIR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PIR. This antibody recognizes human, mouse, rat antigen. The PIR antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
PIR | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
2A10E4 | |
Unconjugated | |
20 μL | |
8544 | |
IgG1 |