Learn More
Hirschmann™ pipetus™ Standard Pump Only 230V UK
Click to view available options
Type:
Pump with Filter
Pump without Filter
Unit Size:
Each
Description
- Spare pump for pipetus™ standard, pump only, 230 Volt UK
Specifications
Specifications
| Type | Pump with Filter |
| For Use With (Equipment) | pipetus™ standard all series |
| Includes | Filter |
| Plug Type | UK |
| Quantity | 1 Each |
| Voltage | 230 V |