Learn More
Nordic Pack Pipettetes
Click to view available options
Volume (Metric):
10 mL
30 mL
50 mL
100 mL
Unit Size:
Each
Description
- Available in different capacities
- Colour: Red
Specifications
Specifications
| Material | Polyethylene |
| Volume (Metric) | 50 mL |
| Type | Dropping Pipette |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.