Learn More
Nordic Pack Pipettetes
Product Code. 11361515
Encompass
Change view
Click to view available options
Volume (Metric):
10 mL
30 mL
50 mL
100 mL
Unit Size:
Each
Product Code. Volume (Metric) unitSize
11361515 30 mL Each
11381515 100 mL Each
11371515 50 mL Each
11351515 10 mL Each
4 options
Product Code. 11361515
Brand: Nordic Pack 1131
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Made from polyethylene
  • Available in different capacities
  • Colour: Red
Specifications
Material Polyethylene
Volume (Metric) 30 mL
Type Dropping Pipette