Learn More
Vitlab™ Pipette Tips
Product Code. 11968091
Change view
Click to view available options
Volume (Metric):
0.5 to 5 mL
1 to 10 mL
Unit Size:
Pack of 200
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11968091 0.5 to 5 mL Pack of 200
11928091 1 to 10 mL Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11968091 Supplier Vitlab™ Supplier No. 146294
  • ONLINE EXCLUSIVE PRICE
    1475.87 NOK
    1244.72 NOK / Pack of 200
    Save 231.15 NOK
    15% Off
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
Vitlab™ Pipette Tips constructed of high-quality polypropylene.
  • Slim shape for pipetting into narrow vessels
  • Made from high-quality polypropylene
  • Bulk tips are delivered in resealable bags to minimize risk of contamination in the lab
Specifications
Description PU 200
Volume (Metric) 0.5 to 5 mL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (Metric) 160mm
Certifications/Compliance CE Marked, IVD 98/79 EC
Diameter (Metric) Outer 9.6mm
Quantity 1 bag with 200 tips