Learn More
BRAND™ Ultra Low Retention Pipette Tips
Significantly reduce sample loss and provide substantially higher reproducibility when working with critical media.
Brand: BRAND™ 732302
Additional Details : Weight : 0.39000kg
Description
- Polypropylene construction
- Cleanroom quality for contamination-free analyses.
- Perfect fit for the Transferpette™ micropipette and many other pipettes.
- Non-sterile
- DNA-, RNase-, endotoxin-free
Specifications
0.1 to 20 mL | |
Autoclavable | |
Autoclave | |
Gray | |
Polypropylene |
Non-sterile | |
CE-IVD, DE-M | |
480/PK | |
Transferpette MicroPipette |