Learn More
Integra Biosciences™ PipetBoy Pro Pipettors
Product Code. 11701258
Click to view available options
Color:
Pink
White
Unit Size:
Each
Product Code. 11701258
Brand: Integra Biosciences™ 156400
Use this pipette controller for precise and efficient serological pipetting. Integra Biosciences™ PipetBoy™ Pro Pipettors feature integrated LED providing an optimal illumination of the area around the pipette for accurate pipetting and preventing eyestrain.
  • Accurately displace liquid in the pipette by applying variable finger pressure on the pipettor triggers
  • Light as a feather, 190 grams
  • Five colors available
  • Speed can be set continuously via the thumb wheel
  • Red LED indicates when the battery needs to be charged
  • Ergonomic design, following the recommendations of hand shape ergonomics
Specifications
Compatible Tips 1 to 100mL Glass and Plastic Pipette
Color White
Dispensing Speed 12 mL/sec.
Model PIPETBOY pro
Type Pipette
Weight (Metric) 190 g