Learn More
Integra Biosciences™ PipetBoy Pro Pipettors
Product Code. 10219874
Change view
Click to view available options
Color:
Pink
White
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10219874 Pink Each
11701258 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10219874 Supplier Integra Biosciences™ Supplier No. 156401
Estimated Shipment: 26-11-2026
Use this pipette controller for precise and efficient serological pipetting. Integra Biosciences™ PipetBoy™ Pro Pipettors feature integrated LED providing an optimal illumination of the area around the pipette for accurate pipetting and preventing eyestrain.
  • Accurately displace liquid in the pipette by applying variable finger pressure on the pipettor triggers
  • Light as a feather, 190 grams
  • Five colors available
  • Speed can be set continuously via the thumb wheel
  • Red LED indicates when the battery needs to be charged
  • Ergonomic design, following the recommendations of hand shape ergonomics
Specifications
Compatible Tips 1 to 100mL Glass and Plastic Pipette
Color Pink
Dispensing Speed 12 mL/sec.
Model PIPETBOY pro
Type Pipette
Weight (Metric) 190 g