Learn More
Vitlab™ Pipet Top
Click to view available options
Type:
Pipette Top
Certifications/Compliance:
DIN EN 285
For Use With (Equipment):
For all Pipettes from 0.1 to 100mL
Autoclavable:
Autoclavable
Unit Size:
Each
Description
- Suction element provides rapid filling of pipettes (capacity: 50 ml in less than 10 seconds)
- Discharge bellows are used for the emptying (blow-out) of the pipette. Moulded intake cone ensures secure seating for all normal bulb and graduated pipettes (0.1 to 200 ml)
- Simple to dismantle, easy to clean
- Autoclavable at 121 °C (2 bar) according to DIN EN 285
- With replacement 3 μm membrane filter and instruction manual
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | DIN EN 285 |
| Quantity | 1 Each |
| Type | Pipette Top |
| For Use With (Equipment) | For all Pipettes from 0.1 to 100mL |