Learn More
BRAND™ Pipette Tips, 2 to 200 μL
GREENER_CHOICE
Product Code. 15104917 Shop All Brand Products
Change view
Click to view available options
Packaging:
Bagged (10,000 Pk.)
Bagged (1000 Pk.)
Tip-Stack (10 x 96 Tips)
TipBox
TipRack
Unit Size:
Pack of 1000
Pack of 480
Pack of 960
Product Code. Packaging unitSize
15104917 TipBox Pack of 480
15154887 Bagged (1000 Pk.) Pack of 1000
15124927 Tip-Stack (10 x 96 Tips) Pack of 960
15134897 Bagged (10,000 Pk.) Pack of 1000
15174897 TipRack Pack of 960
5 options
Product Code. 15104917
Brand: BRAND™ 732208
Only null left
Add to basket
Add to basket
Designed for high-quality micro-pipettes and suitable for various applications. BRAND™ 2 to 200 μL pipette tips ensure precise and efficient pipetting. Tips are cleanroom-produced, free from contaminants, and environmentally friendly.
  • Engineered to fit the Transferpette™ line of micropipettes
  • Compatibility tested with other micropipettors from various manufacturers
  • Manufactured and packaged under certified cleanroom conditions according to ISO 14644-1, Class 7 and 8
  • Made from high-quality, pure raw materials, free of DiHEMDA and oleamide
  • No use of mold release agents such as stearates and erucamide in the production process
  • Palletized BIO-CERT™ quality tips are guaranteed to be free from DNA, DNase, RNase, ATP, and endotoxins
  • Thin-walled for high translucency, easy ejection, and low waste
  • Graduations at 20 and 100 μL for quick volume checks
  • Available either in bulk packaging or convenient, ecological rack-and-stack formats
  • Bulk tips are yellow and placed in clear, resealable bags
  • Racked tips are colorless and placed into yellow-colored tip trays
  • TipBoxes are made of 100% PP and are fully recyclable
  • The TipRack refilling system reduces waste by more than 20% compared to the TipBox
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 2 to 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Colorless
Autoclavable Autoclavable
Clean Claims DNase, RNase, ATP, and Endotoxin Free
Format Box
Tip Style Universal
Packaging TipBox
Product Type Pipette Tips
Disposable Limited-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
Graduations 20, 100 μL