Learn More
Kimble Chase™ Pipet
Specifications
Specifications
| Certifications/Compliance | ASTM E1044 compliant. |
| Description | Fill Volume: 2mL |
| Material | Borosilicate Glass |
| For Use With (Application) | For dispensing viscous liquids and suspensions |
| Graduated | Yes |
| Quantity | 12 Pieces |
| Tip Style | Wide |
| Volume (Metric) | 2 mL |
| Length (Metric) | 350 mm |
| Type | Pipette |