Learn More
Brady™ Pipemarkers on a Card
Click to view available options
Display Text:
Air comprimé
Azote
CO2
Eau chaude sanitaire
Eau déionisée
Eau froide sanitaire
Fuel
Hélium
Color:
Black on Yellow
White on Blue
White on Dark Brown
White on Green
Unit Size:
Pack of 5
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols , if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, which saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
Specifications
Specifications
| Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 |
| Description | Pipe Markers - Fuel |
| Adhesive Type | Permanent Rubber, Permanent, Pressure-sensitive |
| Color | White on Dark Brown |
| Display | Warning |
| Format | Card |
| Material | Polyester |
| Product Type | Pipe Marker |
| Display Text | Fuel |
| For Use With (Application) | Pipe and valve marking |
| Show More |