Learn More
Brady™ Pipemarkers on a Card
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols , if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, which saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
Specifications
Specifications
| Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 |
| Description | Pipe Markers - Eau déionisée |
| Adhesive Type | Permanent Rubber, Permanent, Pressure-sensitive |
| Color | White on Green |
| Format | Card |
| Material | Polyester |
| Product Type | Pipe Marker |
| Display Text | Eau déionisée |
| For Use With (Application) | Pipe and valve marking |
| Labels per Sheet | 5 |
| Show More |