Learn More
ROBU VitraPOR™ Pipeline Filter
Pipeline Filter with tube adapters for connection.
Brand: ROBU VitraPOR™ 23305
Additional Details : Weight : 0.09120kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
Tube Adapters | |
5 | |
30 mm |
Non-sterile | |
2/Pk. | |
140 mm |