missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PIP5K2A (Human) Recombinant Protein
Human PIP5K2A full-length ORF ( AAH18034, 1 a.a. - 406 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005305-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MATPGNLGSSVLASKTKTKKKHFVAQKVKLFRASDPLLSVLMWGVNHSINELSHVQIPVMLMPDDFKAYSKIKVDNHLFNKENMPSHFKFKEYCPMVFRNLRERFGIDDQDFQNSLTRSAPLPNDSQARSGARFHTSYDKRYIIKTITSEDVAEMHNILKKYHQYIVECHGITLLPQFLGMYRLNVDGVEIYVIVTRNVFSHRLSVYRKYDLKGSTVAREASDKEKAKELPTLKDNDFINEGQKIYIDDNNKKVFLEKLKKDVEFLAQLKLMDYSLLVGIHDVERAEQEEVECEENDGEEEGESDGTHPVGTPPDSPGNTLNSSPPLAPGEFDPNIDVYGIKCHENSPRKEVYFMAIIDILTHYDAKKKAAHAAKTVKHGAGAEISTVNPEQYSKRFLDFIGHILT
Specifications
AAH18034 | |
phosphatidylinositol-5-phosphate 4-kinase, type II, alpha | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PIP4K2A | |
GST |
5305 | |
Wheat germ expression system | |
25 μg | |
FLJ13267, PI5P4KA, PIP5K2A, PIP5KII-alpha, PIP5KIIA, PIPK | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |