missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PIP5K1B, GST Tag Recombinant Protein

Product Code. p-7180399
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
20µg
This item is not returnable. View return policy

Product Code. 15940423

Brand: Invitrogen™ A33521

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This PIP5K1B recombinant human protein (full length) was expressed in insect cells. PIP5K1B recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for PIP5K1B is NP_003549.1.

PIP5K1B lipid kinase or phosphatidylinositol-4-phosphate 5-kinase, type I, beta is a member of the phosphatidylinositol-4-phosphate 5-kinase family. PIP5K1B gene contains 17 exons and spans more than 300 kb. The seventeenth exon was found by RT-PCR, and is derived from the 3-prime untranslated region of the PRKACG gene which is located on 9q13 approximately 3 kb downstream of the STM7.I 3-prime untranslated region. The overexpression of PIP5K1B in COS-7 cells induces an increase in short actin fibers and a decrease in actin stress fibers.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14986
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 25% glycerol, 10mM glutathione, 0.25mM DTT, 150mM NaCl, 0.1mM EDTA, 0.1mM PMSF and no preservative; pH 7.5
Gene ID (Entrez) 8395
Molecular Weight (g/mol) 87.3 kDa
Name Human PIP5K1B, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 20 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias MSS4; Phosphatidylinositol 4-phosphate 5-kinase type I alpha; phosphatidylinositol 4-phosphate 5-kinase type I beta; Phosphatidylinositol 4-phosphate 5-kinase type-1 beta; phosphatidylinositol-4-phosphate 5-kinase; phosphatidylinositol-4-phosphate 5-kinase type 1 beta; phosphatidylinositol-4-phosphate 5-kinase type I alpha; phosphatidylinositol-4-phosphate 5-kinase type I beta; phosphatidylinositol-4-phosphate 5-kinase type-1 beta; phosphatidylinositol-4-phosphate 5-kinase, type 1 alpha; phosphatidylinositol-4-phosphate 5-kinase, type 1 beta; phosphatidylinositol-4-phosphate 5-kinase, type 1, beta; phosphatidylinositol-4-phosphate 5-kinase, type I, beta; PI4P5K-I[b]; Pip5k1a; Pip5k1b; PIP5K1-beta; PIP5KIalpha; PIP5KIB2 protein; PIP5KIbeta; Pipk5b; Protein STM-7; ptdIns(4)P-5-kinase 1 beta; ptdIns(4)P-5-kinase beta; STM7; STM-7 protein; Type I phosphatidylinositol 4-phosphate 5-kinase beta; type I phosphatidylinositol-4-phosphate 5-kinase beta
Common Name PIP5K1B
Gene Symbol PIP5K1B
KinaseFamily PI3 Kinase
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.