Learn More
Ohaus™ Pioneer™ PX 224 Analytical Balance
Product Code. 15875327 Shop All Ohaus Products
Change view
Click to view available options
Capacity: 220 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15875327 Capacity: 220 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15875327 Supplier Ohaus™ Supplier No. 30429804
  • ONLINE EXCLUSIVE PRICE
    24175.00 NOK
    16960.97 NOK / Pack of 1
    Save 7214.03 NOK
    29% Off
View all promotions
Estimated Shipment: 02-12-2026
The Pioneer PX combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings.
  • Competitive Performance at an Economical Price - The PX offers high accuracy and repeatability for essential weighing applications in laboratory, industrial and educational settings at an economical pricepoint.
  • Durably Constructed for Long-Term Use - Featuring a cast metal lower housing, sub-pan and stainless steel weighing pan, the PX is durably constructed for versatile, long-term use.
  • Designed for Simple, Intelligent Operation - The PX features a second line display for additional information or guidance, a static removal bar for convenient grounding, and USB and RS232 connectivity for easy communication.
TRUSTED_SUSTAINABILITY
Specifications
Calibration InCal™ - Semi Automatic
Certifications/Compliance CE
Connection Type RS232 & USB
Description Electronic Balance, PX224
Capacity 220 g
Display Type Backlit LCD
Readability 0.000100 g
Languages English (US), French, German, Italian, Spanish (MX)
Linearity 0.000200 g
Material ABS Plastic, Cast Metal, Stainless Steel, Glass
Model PX224
Repeatability 0.000100 g
Stabilization Time 3.000 sec.
Weighing Modes Basic Weighing, Parts Counting, Percent Weighing, Animal Weighing, Density Determination
Dimensions (D x W x H) 321 x 209 x 309 mm
Dimensions (L x W x H) 321.00 x 209.00 x 309.000 mm
Length (Metric) 321.00 mm
Width (Metric) 209.00 mm
Height (Metric) 309.000 mm
Depth (Metric) 321.00 mm
Diameter (Metric) Outer Pan 90 mm
Weight (Metric) 4.5 kg
Weight (Metric) Shipping 7 kg
Electrical Requirements 100/240 V ∼200 mA 50/60 Hz 12/18 VA